Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233176.2 | XP_006233238.1 | 1 - 2 | 253 | 63 | |
XM_006233178.2 | XP_006233240.1 | 1 - 2 | 253 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |