Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044302.1 | NP_001037767.1 | 5 - 6 | 706 | 73 | |
XM_006233176.2 | XP_006233238.1 | 5 - 6 | 916 | 73 | ||
XM_006233177.2 | XP_006233239.1 | 5 - 6 | 747 | 73 | ||
XM_006233178.2 | XP_006233240.1 | 5 - 6 | 916 | 73 | ||
XM_006233179.2 | XP_006233241.1 | 5 - 6 | 747 | 73 | ||
GenBank mRNA | BC105874.1 | - | 5 - 6 | 706 | 7131142 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |