Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085353.1 | NP_001078822.1 | 4 - 5 | 839 | 126 | |
NM_199408.2 | NP_955440.1 | 4 - 5 | 845 | 126 | ||
GenBank mRNA | AY569010.1 | - | 4 - 5 | 845 | 126 | |
BC062391.1 | - | 4 - 5 | 839 | 5621215 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |