Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233347.2 | XP_006233409.1 | 8 - 9 | 4218 | 76 | |
XM_008761534.1 | XP_008759756.1 | 8 - 9 | 4215 | 76 | ||
XM_227694.7 | XP_227694.5 | 7 - 8 | 4101 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |