Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033079.1 | NP_001028251.1 | 5 - 6 | 1224 | 79 | |
GenBank mRNA | L06434.2 | - | 5 - 6 | 1224 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |