Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107717.1 | NP_001101187.1 | 8 - 9 | 1935 | 101 | |
XM_006233159.2 | XP_006233221.1 | 9 - 10 | 1943 | 101 | ||
XM_006233160.2 | XP_006233222.1 | 9 - 10 | 1936 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |