Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761559.1 | XP_008759781.1 | 6 - 7 | 853 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |