Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024157.1 | NP_077071.1 | 6 - 7 | 892 | 84 | |
GenBank mRNA | BC089798.1 | - | 6 - 7 | 897 | 7321559 | 84 |
Y18965.1 | - | 6 - 7 | 892 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |