Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024157.1 | NP_077071.1 | 7 - 8 | 123 | ||
GenBank mRNA | BC089798.1 | - | 7 - 8 | 936 | 7321559 | 123 |
Y18965.1 | - | 7 - 8 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |