Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025688.1 | NP_001020859.1 | 1 - 2 | 295 | 72 | |
GenBank mRNA | BC098755.1 | - | 1 - 2 | 295 | 7370232 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |