Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047918.2 | NP_001041383.2 | 1 - 2 | 72 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |