Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012523.2 | NP_036655.1 | 7 - 8 | 733 | 68 | |
GenBank mRNA | BC078900.1 | - | 7 - 8 | 684 | 7130719 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |