Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023981.4 | NP_076471.3 | 7 - 8 | 1817 | 73 | |
XM_006233187.1 | XP_006233249.1 | 8 - 9 | 1769 | 73 | ||
XM_008761426.1 | XP_008759648.1 | 8 - 9 | 1822 | 73 | ||
XM_008761427.1 | XP_008759649.1 | 9 - 10 | 1744 | 73 | ||
XM_008761428.1 | XP_008759650.1 | 7 - 8 | 1741 | 73 | ||
XM_008761429.1 | XP_008759651.1 | 8 - 9 | 1822 | 73 | ||
GenBank mRNA | AF514355.1 | - | 7 - 8 | 1657 | 73 | |
AF514356.1 | - | 7 - 8 | 1657 | 73 | ||
AF514357.1 | - | 7 - 8 | 1667 | 73 | ||
AF515736.1 | - | 7 - 8 | 1755 | 73 | ||
BC074007.1 | - | 7 - 8 | 1817 | 7099454 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |