Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106475.1 | NP_001099945.1 | 2 - 3 | 235 | 63 | |
XM_006233371.2 | XP_006233433.1 | 3 - 4 | 317 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |