Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133297.2 | NP_579831.2 | 2 - 3 | 525 | 71 | |
GenBank mRNA | AF390544.1 | - | 2 - 3 | 249 | 71 | |
BC060547.1 | - | 2 - 3 | 274 | 6921948 | 71 | |
CK596377.1 | - | 2 - 3 | 540 | 71 | ||
DY561321.1 | - | 2 - 3 | 263 | 71 | ||
FQ212727.1 | - | 2 - 3 | 293 | 71 | ||
FQ213177.1 | - | 2 - 3 | 274 | 71 | ||
FQ213838.1 | - | 2 - 3 | 260 | 71 | ||
FQ214634.1 | - | 2 - 3 | 270 | 71 | ||
FQ215362.1 | - | 2 - 3 | 260 | 71 | ||
FQ215992.1 | - | 2 - 3 | 283 | 71 | ||
FQ216831.1 | - | 2 - 3 | 260 | 71 | ||
FQ219237.1 | - | 2 - 3 | 281 | 71 | ||
FQ219570.1 | - | 2 - 3 | 260 | 71 | ||
FQ219725.1 | - | 2 - 3 | 253 | 71 | ||
FQ220311.1 | - | 2 - 3 | 259 | 71 | ||
FQ220593.1 | - | 2 - 3 | 265 | 71 | ||
FQ220877.1 | - | 2 - 3 | 273 | 71 | ||
FQ229133.1 | - | 2 - 3 | 266 | 71 | ||
FQ230131.1 | - | 2 - 3 | 260 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |