Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191850.1 | NP_001178779.1 | 6 - 7 | 385 | 56 | |
XM_006233291.2 | XP_006233353.2 | 6 - 7 | 718 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |