Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191850.1 | NP_001178779.1 | 7 - 8 | 441 | 121 | |
XM_006233291.2 | XP_006233353.2 | 7 - 8 | 774 | 121 | ||
XM_006233292.2 | XP_006233354.2 | 6 - 7 | 675 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |