Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761474.1 | XP_008759696.1 | 2 - 3 | 139 | 63 | |
GenBank mRNA | FQ210266.1 | - | 2 - 3 | 76 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |