Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135871.1 | NP_001129343.1 | 4 - 5 | 312 | 74 | |
XM_006233327.2 | XP_006233389.1 | 4 - 5 | 1909 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |