Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chi3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191712.1 | NP_001178641.1 | 4 - 5 | 316 | 63 | |
XM_008761383.1 | XP_008759605.1 | 4 - 5 | 316 | 63 | ||
GenBank mRNA | JF781276.1 | - | 4 - 5 | 334 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |