Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039609.1 | NP_001034698.1 | 783 | 159 | ||
GenBank mRNA | AB250295.1 | - | 783 | 159 | ||
AB250296.1 | - | 783 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |