Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173148.1 | NP_775171.1 | 268 | 93 | ||
GenBank mRNA | M76734.1 | - | 268 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |