Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750617.3 | XP_003750665.2 | 11 - 12 | 2708 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |