Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106134.1 | NP_001099604.1 | 1 - 2 | 182 | 139 | |
XM_006254918.2 | XP_006254980.1 | 1 - 2 | 195 | 139 | ||
XM_008772106.1 | XP_008770328.1 | 1 - 2 | 198 | 139 | ||
XM_008772108.1 | XP_008770330.1 | 1 - 2 | 196 | 139 | ||
GenBank mRNA | BC158659.1 | - | 1 - 2 | 134 | 8369180 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |