Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168631.1 | NP_001162102.1 | 8 - 9 | 1642 | 101 | |
XM_006232079.2 | XP_006232141.1 | 8 - 9 | 1718 | 101 | ||
XM_006232080.2 | XP_006232142.1 | 8 - 9 | 1750 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |