Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008770.3 | NP_001008770.1 | 3 - 4 | 361 | 90 | |
XM_006232097.2 | XP_006232159.1 | 3 - 4 | 2472 | 90 | ||
GenBank mRNA | AY325197.1 | - | 3 - 4 | 363 | 90 | |
BC088459.1 | - | 2 - 3 | 345 | 7127481 | 90 | |
BF556107.1 | - | 3 - 4 | 368 | 90 | ||
FQ209709.1 | - | 3 - 4 | 519 | 90 | ||
FQ213936.1 | - | 3 - 4 | 371 | 90 | ||
FQ215832.1 | - | 2 - 3 | 363 | 90 | ||
FQ219291.1 | - | 3 - 4 | 450 | 90 | ||
FQ219760.1 | - | 3 - 4 | 374 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |