Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vma21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771344.1 | XP_008769566.1 | 457 | 85 | ||
GenBank mRNA | FQ215711.1 | - | 452 | 85 |
Gene Symbol: LOC100912478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253887.2 | XP_006253949.1 | 2 - 3 | 457 | 85 |
Gene Symbol: LOC103689964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253890.2 | XP_006253952.1 | 457 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |