Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106808.1 | NP_001100278.1 | 3 - 4 | 368 | 120 | |
XM_008766049.1 | XP_008764271.1 | 2 - 3 | 611 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |