Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108132.1 | NP_001101602.1 | 2 - 3 | 785 | 153 | |
XM_006242720.2 | XP_006242782.1 | 2 - 3 | 780 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |