Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017381.1 | NP_001017381.1 | 1 - 2 | 288 | 75 | |
XM_008766055.1 | XP_008764277.1 | 1 - 2 | 323 | 75 | ||
GenBank mRNA | AY972521.1 | - | 1 - 2 | 288 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |