Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242722.2 | XP_006242784.1 | 20 - 21 | 2151 | 51 | |
XM_235972.8 | XP_235972.6 | 19 - 20 | 2100 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |