Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007661.2 | NP_001007662.1 | 4 - 5 | 435 | 60 | |
GenBank mRNA | BC064031.1 | - | 4 - 5 | 439 | 6887845 | 60 |
FQ213324.1 | - | 3 - 4 | 584 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |