Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108250.1 | NP_001101720.1 | 5 - 6 | 694 | 119 | |
XM_006256731.2 | XP_006256793.1 | 4 - 5 | 757 | 119 | ||
GenBank mRNA | AB232988.1 | - | 5 - 6 | 718 | 119 | |
AB232989.1 | - | 4 - 5 | 636 | 119 | ||
AB232990.1 | - | 5 - 6 | 718 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |