Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012129.2 | NP_001012129.1 | 6 - 7 | 1013 | 87 | |
XM_006244219.2 | XP_006244281.1 | 6 - 7 | 1128 | 87 | ||
XM_006244220.2 | XP_006244282.1 | 6 - 7 | 3042 | 87 | ||
XM_006244221.2 | XP_006244283.1 | 6 - 7 | 1010 | 87 | ||
XM_008766760.1 | XP_008764982.1 | 5 - 6 | 1673 | 87 | ||
XM_008766761.1 | XP_008764983.1 | 6 - 7 | 1642 | 87 | ||
XM_008766762.1 | XP_008764984.1 | 7 - 8 | 1047 | 87 | ||
XM_008766763.1 | XP_008764985.1 | 7 - 8 | 4140 | 87 | ||
XM_008766764.1 | XP_008764986.1 | 5 - 6 | 1673 | 87 | ||
GenBank mRNA | BC085814.1 | - | 6 - 7 | 1014 | 7123102 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |