Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108538.1 | NP_001102008.1 | 9 - 10 | 1286 | 119 | |
XM_008773000.1 | XP_008771222.1 | 11 - 12 | 1256 | 119 | ||
GenBank mRNA | BC166724.1 | - | 11 - 12 | 1217 | 7933637 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |