Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004070.2 | NP_001004070.1 | 1 - 2 | 99 | 105 | |
NM_001165880.1 | NP_001159352.1 | 2 - 3 | 247 | 105 | ||
XM_006255977.2 | XP_006256039.1 | 2 - 3 | 968 | 105 | ||
XM_006255978.2 | XP_006256040.1 | 3 - 4 | 854 | 105 | ||
XM_006255979.2 | XP_006256041.1 | 3 - 4 | 350 | 105 | ||
GenBank mRNA | BC127463.1 | - | 2 - 3 | 247 | 8362353 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |