Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035238.2 | NP_001030315.1 | 5 - 6 | 449 | 72 | |
NM_031021.3 | NP_112283.1 | 5 - 6 | 500 | 72 | ||
XM_006256081.2 | XP_006256143.1 | 5 - 6 | 536 | 72 | ||
GenBank mRNA | BC078807.1 | - | 5 - 6 | 449 | 7133064 | 72 |
EV778558.1 | - | 5 - 6 | 504 | 72 | ||
FQ214437.1 | - | 5 - 6 | 516 | 72 | ||
FQ224491.1 | - | 5 - 6 | 520 | 72 | ||
FQ230013.1 | - | 5 - 6 | 488 | 72 | ||
HE864426.1 | - | 4 - 5 | 384 | 72 | ||
HE864431.1 | - | 384 | 72 | |||
HE864432.1 | - | 384 | 72 | |||
HE864433.1 | - | 384 | 72 | |||
HE864434.1 | - | 384 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |