Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256062.2 | XP_006256124.1 | 1 - 2 | 61 | 69 | |
XM_006256065.2 | XP_006256127.1 | 2 - 3 | 139 | 69 | ||
GenBank mRNA | D82069.1 | - | 2 - 3 | 144 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |