Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212509.2 | NP_997674.1 | 1 - 2 | 168 | 63 | |
GenBank mRNA | BC099772.1 | - | 1 - 2 | 112 | 7932537 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |