Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034157.2 | NP_001029329.1 | 2 - 3 | 788 | 95 | |
XM_006256075.2 | XP_006256137.1 | 2 - 3 | 705 | 95 | ||
XM_006256076.2 | XP_006256138.1 | 2 - 3 | 859 | 95 | ||
XM_006256077.2 | XP_006256139.1 | 3 - 4 | 1112 | 95 | ||
XM_008772761.1 | XP_008770983.1 | 2 - 3 | 829 | 95 | ||
GenBank mRNA | BU758690.1 | - | 1 - 2 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |