Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109137.1 | NP_001102607.1 | 4 - 5 | 722 | 93 | |
XM_006227887.2 | XP_006227949.1 | 5 - 6 | 3075 | 93 | ||
XM_008758736.1 | XP_008756958.1 | 5 - 6 | 2559 | 93 | ||
XM_008758737.1 | XP_008756959.1 | 5 - 6 | 3247 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |