Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751734.3 | XP_003751782.2 | 1 - 2 | 1245 | 148 | |
GenBank mRNA | BC158695.1 | - | 1 - 2 | 125 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |