Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751734.3 | XP_003751782.2 | 5 - 6 | 1704 | 100 | |
XM_008771923.1 | XP_008770145.1 | 5 - 6 | 1744 | 100 | ||
GenBank mRNA | BC158695.1 | - | 5 - 6 | 584 | 100 | |
FQ220359.1 | - | 2 - 3 | 174 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |