Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106629.1 | NP_001100099.1 | 2 - 3 | 173 | 101 | |
GenBank mRNA | FQ220333.1 | - | 2 - 3 | 178 | 101 | |
FQ220437.1 | - | 2 - 3 | 189 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |