Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106629.1 | NP_001100099.1 | 3 - 4 | 687 | 60 | |
GenBank mRNA | FQ220333.1 | - | 3 - 4 | 692 | 60 | |
FQ220437.1 | - | 3 - 4 | 703 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |