Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031987.1 | NP_114193.1 | 13 - 14 | 1474 | 105 | |
XM_006236004.2 | XP_006236066.1 | 13 - 14 | 1634 | 105 | ||
XM_006236005.2 | XP_006236067.1 | 14 - 15 | 2964 | 105 | ||
GenBank mRNA | BC074004.1 | - | 13 - 14 | 1494 | 7098081 | 105 |
J02844.1 | - | 1914 | 105 | |||
U26033.1 | - | 13 - 14 | 1474 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |