Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ011608.1 | - | 5 - 6 | 592 | 108 | |
BC088272.1 | - | 5 - 6 | 599 | 7379940 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |