Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198562.1 | NP_001185491.1 | 1 - 2 | 69 | 80 | |
GenBank mRNA | BC166934.1 | - | 1 - 2 | 57 | 6922001 | 80 |
BP494723.1 | - | 1 - 2 | 91 | 80 | ||
FQ221678.1 | - | 1 - 2 | 102 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |