Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031086.2 | NP_112348.2 | 13 - 14 | 1725 | 86 | |
XM_006240719.2 | XP_006240781.1 | 11 - 12 | 2011 | 86 | ||
XM_008765045.1 | XP_008763267.1 | 11 - 12 | 1457 | 86 | ||
GenBank mRNA | CK366505.1 | - | 1 - 2 | 150 | 86 | |
CV105055.1 | - | 4 - 5 | 680 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |