Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106209.1 | NP_001099679.1 | 2 - 3 | 577 | 73 | |
XM_006227918.2 | XP_006227980.1 | 4 - 5 | 1477 | 73 | ||
XM_008758750.1 | XP_008756972.1 | 1 - 2 | 726 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |